All Photos(1)
About This Item
Linear Formula:
Cl2C6F4
CAS Number:
Molecular Weight:
218.96
EC Number:
MDL number:
UNSPSC Code:
12352100
Assay
85%
refractive index
n20/D 1.468 (lit.)
bp
150-152 °C (lit.)
density
1.652 g/mL at 25 °C (lit.)
SMILES string
Fc1c(F)c(Cl)c(F)c(Cl)c1F
Other Notes
remainder 1,2-isomer
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service