Skip to Content
Merck
All Photos(1)

Documents

339679

Sigma-Aldrich

3′-Bromo-4′-fluoroacetophenone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)COCH3
CAS Number:
Molecular Weight:
217.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

52-57 °C (lit.)

SMILES string

CC(=O)c1ccc(F)c(Br)c1

InChI

1S/C8H6BrFO/c1-5(11)6-2-3-8(10)7(9)4-6/h2-4H,1H3

InChI key

SZDWTGAORQQQGY-UHFFFAOYSA-N

General description

3′-Bromo-4′-fluoroacetophenone is also referred as 3-bromo-4-fluoro-acetophenone. Palladium-catalyzed amination of 3-bromo-4-fluoro-acetophenone has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed amination of 3-bromo-4-fluoro-acetophenone.
Sun L-Q, et al.
Tetrahedron Letters, 43(51), 9291-9294 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service