Skip to Content
Merck
All Photos(1)

Key Documents

326100

Sigma-Aldrich

Ammonium hexachloroplatinate(IV)

Synonym(s):

Ammonium platinic chloride, Platinum(IV)-ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2PtCl6
CAS Number:
Molecular Weight:
443.87
EC Number:
MDL number:
UNSPSC Code:
12352302

density

3.07 g/mL at 25 °C (lit.)

SMILES string

[NH4+].[NH4+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2H3N.Pt/h6*1H;2*1H3;/q;;;;;;;;+4/p-4

InChI key

PCCGQTHFYHJATL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Reacts with Cu(OAc)2 and H2S to form Cu2PtIIPt3IVS8 which has applications in diamagnetic materials and semiconductors.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service