Skip to Content
Merck
All Photos(1)

Key Documents

214558

Acid Red 40

Dye content 40 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17N3Na2O8S2
CAS Number:
Molecular Weight:
537.47
Colour Index Number:
18070
MDL number:

composition

Dye content, 40%

SMILES string

[Na+].[Na+].CC(=O)Nc1cc(cc2cc(c(\N=N\c3cc(C)cc(C)c3)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service