Skip to Content
Merck
All Photos(1)

Key Documents

184306

Sigma-Aldrich

3-Methoxy-4-nitrobenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
197.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

233-235 °C (lit.)

SMILES string

COc1cc(ccc1[N+]([O-])=O)C(O)=O

InChI

1S/C8H7NO5/c1-14-7-4-5(8(10)11)2-3-6(7)9(12)13/h2-4H,1H3,(H,10,11)

InChI key

PWURRRRGLCVBMX-UHFFFAOYSA-N

Application

3-Methoxy-4-nitrobenzoic acid was used in detection of nitroaromatic compounds by CdSe- quantum dots capped with polyamidoamine dendrimer-1,12-diaminododecane core-generation 4 nanocomposites. It was used in the synthesis of vorozole derivatives.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul M Wood et al.
ChemMedChem, 6(8), 1423-1438 (2011-05-25)
Concurrent inhibition of aromatase and steroid sulfatase (STS) may provide a more effective treatment for hormone-dependent breast cancer than monotherapy against individual enzymes, and several dual aromatase-sulfatase inhibitors (DASIs) have been reported. Three aromatase inhibitors with sub-nanomolar potency, better than
M Algarra et al.
Talanta, 83(5), 1335-1340 (2011-01-18)
The detection of nitroaromatic compounds, best known as raw materials in explosives preparations, is important in many fields including environmental science, public security and forensics. CdSe quantum dots capped with PAMAM-G(4) dendrimer were synthetized in water and used for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service