182109
Poly(hexadecyl methacrylate) solution
average Mw ~200,000 by GPC, in toluene
Synonym(s):
Cetyl methacrylate homopolymer, Hexadecyl methacrylate homopolymer, Palmityl methacrylate homopolymer, Poly(cetyl methacrylate)
About This Item
Recommended Products
form
liquid
Quality Level
mol wt
average Mw ~200,000 by GPC
concentration
in toluene
transition temp
Tg 15 °C
SMILES string
CCCCCCCCCCCCCCCCOC(=O)C(C)=C
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
39.2 °F - closed cup
Flash Point(C)
4 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service