Skip to Content
Merck
All Photos(1)

Key Documents

13900

Sigma-Aldrich

O-Benzyl-L-serine

≥99.0% (NT)

Synonym(s):

(S)-2-Amino-3-benzyloxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO3
CAS Number:
Molecular Weight:
195.22
Beilstein:
2114846
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (NT)

form

powder

optical activity

[α]20/D +21±2°, c = 2% in acetic acid: water (4:1) (+ 1 Eq HCl)

optical purity

enantiomeric ratio: ≥99.5:0.5 (TLC)

reaction suitability

reaction type: solution phase peptide synthesis

mp

~227 °C (dec.)

application(s)

peptide synthesis

SMILES string

N[C@@H](COCc1ccccc1)C(O)=O

InChI

1S/C10H13NO3/c11-9(10(12)13)7-14-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)/t9-/m0/s1

InChI key

IDGQXGPQOGUGIX-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amir H Ghassemi et al.
Pharmaceutical research, 27(9), 2008-2017 (2010-07-06)
To study the release of a model protein, bovine serum albumin (BSA), from microspheres of an hydroxylated aliphatic polyester, poly(lactic-co-hydroxymethyl glycolic acid) (PLHMGA). BSA-loaded microspheres were prepared by a double emulsion solvent evaporation method. The effect of copolymer composition and
Amir H Ghassemi et al.
Pharmaceutical research, 29(1), 110-120 (2011-07-12)
To investigate the in vitro release of octreotide acetate, a somatostatin agonist, from microspheres based on a hydrophilic polyester, poly(D,L-lactide-co-hydroxymethyl glycolide) (PLHMGA). Spherical and non-porous octreotide-loaded PLHMGA microspheres (12 to 16 μm) and loading efficiency of 60-70% were prepared by a
Roman V Uzhachenko et al.
Cell reports, 35(1), 108944-108944 (2021-04-08)
Inhibitors of cyclin-dependent kinases 4 and 6 (CDK4/6i) delay progression of metastatic breast cancer. However, complete responses are uncommon and tumors eventually relapse. Here, we show that CDK4/6i can enhance efficacy of T cell-based therapies, such as adoptive T cell transfer or
Neda Samadi et al.
Pharmaceutical research, 31(10), 2593-2604 (2014-03-15)
To investigate the effect of polyethylene glycol (PEG) in nanoparticles based on blends of hydroxylated aliphatic polyester, poly(D,L-lactic-co-glycolic-co-hydroxymethyl glycolic acid) (PLGHMGA) and PEG-PLGHMGA block copolymers on their degradation and release behavior. Protein-loaded nanoparticles were prepared with blends of varying ratios
F Kazazi-Hyseni et al.
International journal of pharmaceutics, 482(1-2), 99-109 (2014-12-17)
Poly(D,L-lactic-co-hydroxymethyl glycolic acid) (PLHMGA) is a biodegradable copolymer with potential as a novel carrier in polymeric drug delivery systems. In this study, the biocompatibility of PLHMGA microspheres (PLHMGA-ms) was investigated both in vitro in three different cell types (PK-84, HK-2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service