AC5410537750
Accela ChemBio Inc.
Methyl 5-(3-Pyridyl)isoxazole-3-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 5-(3-pyridinyl)-3-isoxazolecarboxylate
Inchi Code
1S/C10H8N2O3/c1-14-10(13)8-5-9(15-12-8)7-3-2-4-11-6-7/h2-6H,1H3
InChI key
IJYBKCVFGWEEKG-UHFFFAOYSA-N
storage temp.
+4C
Purity
97%
Country of Origin
CN
Shipping Temperature
RT
Physical Form
White to brown solid
Product Link
http://www.accelachem.com/cn/search.html?keyword=SY015172&attname=GoodsCode
MSDS
http://www.accelachem.com/cn/msds/SY015172.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.