FAP88987
Pesticides and PCBs in infant formula
reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
reference material
Quality Level
manufacturer/tradename
Fera Science Ltd
application(s)
food and beverages
format
matrix material
storage temp.
2-8°C
Related Categories
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants / Pesticides
Chlordane (cis), Chlorfenvinphos (sum of E and Z isomers), HCH-B (beta hexachlorocyclohexane), Heptachlo-eopxide (cis), PCB 101, Demeton-S-methyl-sulfoxide (oxydemeton-methyl), Nitrofen
Matrix Group: Dairy products and eggs
Organic Pollutants / Pesticides
Chlordane (cis), Chlorfenvinphos (sum of E and Z isomers), HCH-B (beta hexachlorocyclohexane), Heptachlo-eopxide (cis), PCB 101, Demeton-S-methyl-sulfoxide (oxydemeton-methyl), Nitrofen
Matrix Group: Dairy products and eggs
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service