Skip to Content
Merck
All Photos(2)

Key Documents

SRP0706

Sigma-Aldrich

KAT5 (TIP60), Active

recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

25-35 nmol/min·mg (radiometric)

mol wt

~88 kDa

concentration

0.1 μg/μL

accession no.

NM_006388

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KAT5(10524)

Application

Useful for acetyltransferase assay.

Biochem/physiol Actions

Full-length recombinant human KAT5 was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. KAT5 or K (lysine) acetyltransferase 5 is a signaling protein that belongs to the MYST family of histone acetyl transferases (HATs). KAT5 was originally isolated as an HIV-1 TAT-interactive protein that plays an important role in regulating chromatin remodeling, transcription and other nuclear processes by acetylating histone and nonhistone proteins (1). KAT5 protein is a histone acetylase that is involved in DNA repair and apoptosis and plays an important role in signal transduction. KAT5 acts as a cofactor of TAT that is involved in the regulation of HIV gene expression. KAT5 is also a co-regulator of transcription factors that either promote or suppress tumorigenesis (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service