Skip to Content
Merck
All Photos(1)

Documents

MAK408

Sigma-Aldrich

Phosphoenolpyruvate Carboxykinase Activity Assay Kit (Colorimetric)

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes

storage temp.

−20°C

General description

Phosphoenolpyruvate Carboxykinase (PEPCK) is an enzyme which belongs to the lyase family. In the presence of GTP, it catalyzes the reversible conversion of oxaloacetate (OAA) into phosphoenolpyruvate (PEP), GDP and CO2. In humans, two isoforms of PEPCK are found: cytosolic form (PEPCK-C, also called PCK1) and mitochondrial form (PEPCK-M). PEPCK-C is a rate-controlling step in gluconeogenesis. Recent studies found abnormal concentrations of PEPCK in diabetic mice. Therefore, accurate measurement of Phosphoenolpyruvate Carboxykinase activity is valuable for both mechanistic and therapeutic studies.

Features and Benefits

Phosphoenolpyruvate Carboxykinase Activity Assay Kit:
  • Compatible with high-throughput handling systems.
  • Sensitive, detecting than 10 μU of Phosphoenolpyruvate Carboxykinase activity per sample.

Suitability

The kit is suitable for the measurement of Phosphoenolpyruvate Carboxykinase activity in various samples including tissue (rat liver, kidney, heart etc.) and adherent/suspension cells (HeLa, Jurkat, HEK 293 cells etc.). As well as the analysis of the gluconeogenesis pathway.

Principle

In this assay, Phosphoenolpyruvate Carboxykinase is coupled with a set of enzymes that convert phosphoenolpyruvate and carbonate into a series of intermediates and hydrogen peroxide, which in turn, reacts with a probe and is converted generating a colorimetric signal (A570 nm). The color intensity is directly proportional to the amount of active Phosphoenolpyruvate Carboxykinase present in samples.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service