Skip to Content
Merck
All Photos(1)

Key Documents

HPA009027

Sigma-Aldrich

Anti-ULK2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Serine/threonine-protein kinase ULK2 antibody produced in rabbit, Anti-Unc-51-like kinase 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20-1:50

immunogen sequence

VVRRSNTSPMGFLRPGSCSPVPADTAQTVGRRLSTGSSRPYSPSPLVGTIPEQFSQCCCGHPQGHDSRSRNSSGSPVPQAQSPQSLLSGARLQSAPTLTDIYQNKQKLRKQHSDPVCPSHTGAGYSYSPQPSRPGSLGTSPTKH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ULK2(9706)

General description

ULK2 (unc-51 like autophagy activating kinase 2) is an autophagy inducer gene. This protein is a mammalian homolog of the yeast Saccharomyces cerevisiae protein autophagy-related proteins (ATGs). It is a Ser/Thr kinase.

Immunogen

Serine/threonine-protein kinase ULK2 recombinant protein epitope signature tag (PrEST)

Application

Anti-ULK2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ULK2 (unc-51 like autophagy activating kinase 2) is essential for the induction of autophagy, and is involved in multiple physiological, developmental and pathological processes. ULK2 and ULK1 form complexes with Atg13 and FIP200, which are direct targets of mammalian target of rapamycin (mTOR). Thus, they control autophagy in an mTOR-dependent manner. It is hypermethylated and hence, down-regulated in glioblastoma (GBM) and glioma cell lines. This can be reversed by methylation inhibitor treatment. Donw-regulation of this protein results in suppressed autophagy, which is essential for the development of glioma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71405

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sudhanshu Shukla et al.
The Journal of biological chemistry, 289(32), 22306-22318 (2014-06-14)
Glioblastoma (GBM) is the most aggressive type of brain tumor and shows very poor prognosis. Here, using genome-wide methylation analysis, we show that G-CIMP+ and G-CIMP-subtypes enrich distinct classes of biological processes. One of the hypermethylated genes in GBM, ULK2
Chang Hwa Jung et al.
Molecular biology of the cell, 20(7), 1992-2003 (2009-02-20)
Autophagy, the starvation-induced degradation of bulky cytosolic components, is up-regulated in mammalian cells when nutrient supplies are limited. Although mammalian target of rapamycin (mTOR) is known as the key regulator of autophagy induction, the mechanism by which mTOR regulates autophagy
W Gao et al.
Cell death and differentiation, 18(10), 1598-1607 (2011-04-09)
In yeast, activation of ATG1/ATG13 kinase complex initiates autophagy. This mechanism of autophagy initiation is conserved, as unc-51-like kinase 1 (ULK1) and unc-51-like kinase 2 (ULK2) are two mammalian functional homologues of ATG1 and form similar complex with mammalian ATG13.
Rushika M Perera et al.
Nature, 524(7565), 361-365 (2015-07-15)
Activation of cellular stress response pathways to maintain metabolic homeostasis is emerging as a critical growth and survival mechanism in many cancers. The pathogenesis of pancreatic ductal adenocarcinoma (PDA) requires high levels of autophagy, a conserved self-degradative process. However, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service