BMSIPD960035PTFE
BeadBlaster™ 96 Grinding Jars for Pharmaceutical Samples
PTFE, capacity 35 mL, pack of 1
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
Recommended Products
material
PTFE
packaging
pack of 1
manufacturer/tradename
Benchmark Scientific IPD9600-35PTFE
capacity
35 mL
compatibility
BeadBlaster™ 96
Related Categories
General description
Samples require a jar and grinding balls that are just as hard or harder for effective sample preparation. All of the jars feature threaded closures for leakproof performance with both solids and liquids. PTFE is the least hard of the materials used in the jars and is chosen for its inert properties.
PTFE jars are metal-free and non-stick. They are highly temperature resistant, expanding their use to cryogrinding. PTFE jars are used for applications in the pharmaceutical field as well as for plants and other biological samples. The 15mm PTFE grinding balls have steel cores for a higher energy input. The jars are supplied as a 35ml volume
PTFE jars are metal-free and non-stick. They are highly temperature resistant, expanding their use to cryogrinding. PTFE jars are used for applications in the pharmaceutical field as well as for plants and other biological samples. The 15mm PTFE grinding balls have steel cores for a higher energy input. The jars are supplied as a 35ml volume
Application
Homogenization of Pharmaceutical samples, cryogrinding
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)