33-036
PI 3-Kinase HTRF® Assay; 10,000 wells
The PI 3-Kinase HTRF Assay is a high-performance assay kit that provides a universal method for testing all Class I PI3-Kinases in a homogeneous format. This pack size has a screening capacity of 10,000 wells.
About This Item
Recommended Products
Quality Level
species reactivity
mouse, human
manufacturer/tradename
HTRF®
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
detection method
fluorometric (HTRF)
shipped in
dry ice
Gene Information
human ... PIK3CA(5290)
General description
Application
Biochem/physiol Actions
Packaging
Components
PIP2 (Substrate; 1mM stock)
Stop Reagent A
Stop Reagent B
DM A Detection Reagent
DM B Detection Reagent
DM C Detection Reagent
Storage and Stability
Legal Information
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service