NIOSH Method 7903: IC Analysis of Inorganic Acids after Collection/Desorption using ORBO™-53
Materials
related product
Product No.
Description
Pricing
ORBO™ 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters
GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 eaCONDITIONS
sample/matrix
1 ppm of each component
adsorbent tube
ORBO-53 (20265)
column
Dionex S2 Fast Run Anion, 25 cm × 3 mm I.D.
mobile phase
3 mM sodium bicarbonate; 2.4 mM sodium carbonate
flow rate
3 mL/min
detector
conductivity (10μSiemans/ cm)
Description
Legal Information
ORBO is a trademark of Sigma-Aldrich Co. LLC