Skip to Content
Merck
All Photos(1)

Key Documents

R1513

Sigma-Aldrich

Cryogenic storage rack, stainless steel

Horizontal; 16-shelf for 51/4 × 51/4 × 2 in. boxes

Synonym(s):

cryogenic vial rack, cryostorage rack, cryovial rack, dewar rack, liquid nitrogen rack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122806
NACRES:
NB.22
Pricing and availability is not currently available.

material

stainless steel

feature

Holds 16 x boxes, 5¼ × 5 ¼ × 2 in

packaging

pack of 2 ea

manufacturer/tradename

Nalgene® 5038-4422

technique(s)

cryopreservation: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Racks hold CryoBoxes securely but separately, allowing fast retrieval and immediate return to freezer. The vertical racks are best for use in chest freezers, horizontal racks for upright freezers.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service