Skip to Content
Merck
All Photos(1)

Key Documents

1.46631

Millipore

Fluid Thioglycollate Medium

bottle capacity 500 mL, bottle filling volume 500 mL, closure type, Red flip cap with septum, pack of 6 bottles

Synonym(s):

FTM, Fluid Thioglycollate Medium, Thioglycollate Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type Red flip cap with septum
ready-to-use

packaging

pack of 6 bottles

bottle capacity

500 mL

bottle filling volume

500 mL

pH

7.1±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

components

tween: 0.1%

storage temp.

2-25°C

suitability

nonselective for

General description

Fluid Thioglycollate Medium also known as FTM is a nutrient rich complex medium designed to serve as a sterility control for pharmaceutical products.
The presence of Thioglycollate and L-Cystine in the medium reduces the redox potential of the medium to create an anaerobic atmosphere. Additionally, it deactivates the presence of mercury and other heavy metals.
The presence of agar further reduces oxygen diffusion and resazurin indicates the reduced oxygen potential in the medium. An increased oxygen concentration will render a color change from yellow to pink in the medium.

Application

Fluid Thioglycollate Medium is used to isolate and cultivate fastidious anaerobic and aerobic microbes in test samples. FTM is suitable as a sterility control of pharmaceutical products.
Fluid Thioglycollate Medium is recommended for sterility testing of samples according to EU and US pharmacopeia.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They are formulated and tested to meet the pharmacopeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, thus improving accuracy, and reducing the risk of incorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service