Skip to Content
Merck
All Photos(1)

Key Documents

05-777-AF647

Sigma-Aldrich

Anti-Phosphotyrosine Antibody, recombinant clone 4G10® Antibody, Alexa Fluor 647

clone 4G10, from mouse, ALEXA FLUOR 647

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
conjugate:
ALEXA FLUOR 647
application:
ICC
clone:
4G10, monoclonal
technique(s):
immunocytochemistry: suitable
citations:
1

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G10, monoclonal

species reactivity (predicted by homology)

all

technique(s)

immunocytochemistry: suitable

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr)

General description

Phosphorylation plays an important role in regulating protein activities and various cellular signaling events. Posttranslational phosphorylation can occur on histidine (pHis), serine (pSer), threonine (pThr), and tyrosine (pTyr) residues. Due to the early advent of anti-pTyr antibodies, tyrosine phosphorylation remains the mostly studied signaling events in all biological research fields. Tyrosine phosphorylation is considered to be one of the key steps in signal transduction and regulation of enzymatic activity. Before the availability of anti-pTyr antibodies, protein tyrosine phosphorylation was routiniely detected by time-consuming radioactive labeling. Although radioactive labeling is still considered as more sensitive, advancement in detection technology has greatly improved the detection limits of antibody-based methods. Anti-pTyr antibodies are commonly used in ELISA, immunoprecipitation, immunoblotting (including dot blot and Western blot), as well as indirect immunofluourescent detection of tyrosine phosphorylation in tissue and cell samples.

Specificity

Recognizes tyrosine-phosphorylated proteins from all species.
Target modification is not species-specific

Immunogen

Produced from CHO cells expressing the 4G10 antibody heavy and light chain cDNAs. Purified via heavy chain C-terminus hexa-histidine tag by Nickel affinity matrices prior to conjugation with Alexa Fluor 647.

Application

Anti-Phosphotyrosine Antibody, clone 4G10, Alexa Fluor 647 is a highly validated recombinant mouse monoclonal antibody that targets protein tyrosine phosphorylation and has been tested in Immunocytochemistry.
Research Category
Signaling
This recombinant monoclonal antibody is also available as HRP conjugate (Cat. No. 16-184), agarose conjugate (Cat. No. 16-199), biotin conjugate (Cat. No. 16-204), FITC conjugate (Cat. No. 16-205), PE conjugate (Cat. No. FCMAB323PE), as well as as unconjugated antibody (Cat. No. 05-777) for immunoaffinity purification, flow cytometry, immunocytochemistry, immunoprecipitation, and Western blotting applications.

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected EGF-induced tyrosine phosphorylation of cellular proteins in A431 cells.

Target description

Variable depending on the size(s) of the tyrosine-phosphorylated protein(s).

Physical form

Protein G purified.
Purified mouse monoclonal IgG2a antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anees Ahmad Banday
American journal of physiology. Renal physiology, 311(5), F958-F966 (2016-11-03)
The regulation of Na-K-ATPase in various tissues is under the control of a number of hormones and peptides that exert both short- and long-term control over its activity. The present study was performed to investigate the effect of chronic insulin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service