Key Documents
Z176052
Sigma-Aldrich
Lecture-bottle valve
CGA Inlet for 180M/110F, 316 stainless steel
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
material
316 stainless steel
fitting
CGA Inlet for 180M/110F
General description
Diffusion resistant, diaphragm seal, packless on/off valve. 3/8 in. NPTM valve thread. CGA 180M inlet/110F outlet connection.
Choose from one of the most recent versions:
Certificates of Analysis (COA)