822876
TOYOPEARL GigaCap® S-650S Bulk Media
bottle of 250 mL, phase sulphonyl, 35 μm particle size
Synonym(s):
TOYOPEARL GigaCap® S-650 Cation Exchange Media
About This Item
product line
TOYOPEARL®
form
slurry
feature
counter ion Na+
packaging
bottle of 250 mL
manufacturer/tradename
Tosoh 22876
parameter
3 bar max. pressure
technique(s)
LPLC: suitable
matrix
hydroxylated methacrylic polymer
matrix active group
sulphonyl phase
particle size
35 μm
operating pH
3-13
capacity
156 g/L±20 adsorption capacity (hIgG)
separation technique
cation exchange
Looking for similar products? Visit Product Comparison Guide
General description
While Toyopearl GigaCap S-650M resin was specifically optimized for IgG, with dynamic binding capacities in the vicinity of 140 mg/mL for IgG, it also exhibits high binding capacities for smaller proteins. Typical dynamic binding capacities for lysozyme are in excess of 160 mg/mL.
Toyopearl GigaCap S-650S resin has dynamic binding capacities approaching 165 g/L for gamma globulin. This high capacity, combined with the 35 μm mean particle size, makes Toyopearl GigaCap S-650S ideal for high resolution applications.
Toyopearl GigaCap S-650 resin is stable at high linear velocities, has good pressure-flow characteristics and will withstand back pressures up to 3 bar. The higher capacity and back pressure stability of Toyopearl GigaCap S-650 resin creates opportunities for increased throughput in downstream cation exchange purification steps.
Toyopearl GigaCap S-650M resin also has excellent base stability in 1.0 mol/L NaOH and shows robust clean in place (CIP) properties with minimal loss of capacity after 50 X 1 hour cycles of caustic exposure also using 1.0 mol/L NaOH. (data)
*specifically designed for packed bed use
Specifications
Physical form
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
104.0 °F - closed cup
Flash Point(C)
40 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service