Skip to Content
Merck
All Photos(1)

Documents

56664-U

Supelco

Micro-Mixer Static Mixers (2 - 25 μL)

25 μL cartridge, stainless steel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

material

stainless steel

compatibility

for use with ASI Micro Mixer Housing

Looking for similar products? Visit Product Comparison Guide

General description

Micro-Mixer Cartridge, of 25 μL volume, is made of stainless steel material, to match the plumbing in your pump or delivery system and chemical sensitivity. It is used along with ASI Micro Mixer housing (In-line, Binary, Ternary), in ASI Static Mixers for HPLC and post-column derivatization.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service