SML1578
Aftin-4
≥98% (HPLC)
Synonym(s):
(2R)-2-[[9-(1-Methylethyl)-6-[methyl(phenylmethyl)amino]-9H-purin-2-yl]amino]-1-butanol, Amyloid-β Forty-Two Inducer 4, N6-Methyl-(R)-roscovitine
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
storage condition
protect from light
color
white to beige
solubility
DMSO: 25 mg/mL, clear
storage temp.
−20°C
SMILES string
[n]1(c2nc(nc(c2nc1)N(Cc3ccccc3)C)N[C@@H](CO)CC)C(C)C
InChI
1S/C20H28N6O/c1-5-16(12-27)22-20-23-18(25(4)11-15-9-7-6-8-10-15)17-19(24-20)26(13-21-17)14(2)3/h6-10,13-14,16,27H,5,11-12H2,1-4H3,(H,22,23,24)/t16-/m1/s1
InChI key
YPYWONAECUVKHY-MRXNPFEDSA-N
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service