Skip to Content
Merck
All Photos(1)

Key Documents

MABT1364

Sigma-Aldrich

Anti-Spectrin beta-II Antibody, clone 2A7

clone 2A7, from mouse

Synonym(s):

Spectrin beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
clone:
2A7, monoclonal
application:
ICC
IP
WB
species reactivity:
human, rabbit
technique(s):
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A7, monoclonal

species reactivity

human, rabbit

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SPTBN1(6711)

General description

Spectrin beta (UniProt: G1SN11) is encoded by the SPTBN1 gene in rabbit. Spectrins belong to the superfamily of proteins called F-actin cross linking proteins that function as scaffolding proteins for protein sorting, cell adhesion, and migration. They are principle components of a cell membrane-cytoskeleton and are composed of two alpha and two beta spectrin subunits. They are actin binding proteins that serves as a membrane organizer and stabilizer. Spectrin beta II is a multifunctional protein that contains lipid-binding sites within its two calponin-homology domains (aa 82-186 and 201-306) and a pleckstrin homology domain (aa 2224-2334) and triple helical segments. Spectrin beta II is generally associated with the cytoplasmic surface of the membrane by attachment to ankyrin.

Specificity

Clone 2A7 is a mouse monoclonal antibody that detects beta-II spectrin.

Immunogen

Purified spectrin from frozen rabbit lens membranes.

Application

Anti-Spectrin beta-II , clone 2A7, Cat. No. MABT1364, is a mouse monoclonal antibody that detects Spectrin beta chain and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: A representative lot immunoprecipitated Spectrin beta-II in Immunoprecipitation applications (Bazellieres, E., et. al. (2012). J Cell Sci. 125(Pt 4):919-31).

Immunocytochemistry Analysis: A representative lot detected Spectrin beta-II in Immunocytochemistry applications (Bazellieres, E., et. al. (2012). J Cell Sci. 125(Pt 4):919-31).

Western Blotting Analysis: A representative lot detected Spectrin beta-II in Western Blotting applications (Bazellieres, E., et. al. (2012). J Cell Sci. 125(Pt 4):919-31).
Research Category
Cell Structure

Quality

Evaluated by Western Blotting in differentiated Caco2 cell lysates.

Western Blotting Analysis: 2 µg/mL of this antibody detected Spectrin beta-II in differentiated Caco2 cell lysates.

Target description

~275 kDa observed; 271.27 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service