Skip to Content
Merck
All Photos(1)

Key Documents

8.08177

Sigma-Aldrich

Dimethyl terephthalate

for synthesis

Synonym(s):

Dimethyl terephthalate, Dimethyl 1,4-benzenedicarboxylate, Terephthalic acid dimethyl ester, DMT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(COOCH3)2
CAS Number:
Molecular Weight:
194.18
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-411-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

autoignition temp.

520 °C (DIN 51794)

potency

3200 mg/kg LD50, oral (Rat)

expl. lim.

0.8-11.8 % (v/v)

bp

282 °C/1013 hPa

mp

139-141 °C

transition temp

flash point 154 °C (External MSDS)

density

1.35 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

O(C)C(=O)c1ccc(cc1)C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h3-6H,1-2H3

InChI key

WOZVHXUHUFLZGK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 139 °C
Melting range (upper value): ≤ 142 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

309.2 °F - (External MSDS)

Flash Point(C)

154 °C - (External MSDS)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service