8.01272
2-Aminothiazole
for synthesis
Synonym(s):
2-Aminothiazole, 2-Thiazolylamine
About This Item
Recommended Products
vapor pressure
<1 hPa ( 20 °C)
Quality Level
Assay
≥98.0% (GC)
form
powder
autoignition temp.
600 °C
potency
480 mg/kg LD50, oral (Rat)
pH
9.6 (20 °C, 100 g/L in H2O)
bp
117 °C/20 hPa
mp
86-91 °C
solubility
100 g/L
bulk density
600 kg/m3
storage temp.
2-30°C
InChI
1S/C3H4N2S/c4-3-5-1-2-6-3/h1-2H,(H2,4,5)
InChI key
RAIPHJJURHTUIC-UHFFFAOYSA-N
Analysis Note
Melting range (lower value): ≥ 86 °C
Melting range (upper value): ≤ 91 °C
Identity (IR): passes test
Chloride (Cl): ≤ 0.50 %
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service