646121
4-Methoxytriphenylamine
97%
Synonym(s):
4-Methoxy-N,N-diphenylaniline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H17NO
CAS Number:
Molecular Weight:
275.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
102-106 °C (lit.)
SMILES string
COc1ccc(cc1)N(c2ccccc2)c3ccccc3
InChI
1S/C19H17NO/c1-21-19-14-12-18(13-15-19)20(16-8-4-2-5-9-16)17-10-6-3-7-11-17/h2-15H,1H3
InChI key
KIGTXAWIOISJOG-UHFFFAOYSA-N
Application
4-Methoxytriphenylamine (P-MOTPA) can be used as a monomer precursor for the preparation of poly-4-methoxytriphenylamine via oxidative polymerization using FeCl3 as an oxidant. Mixture of P-MOTPA and N,N′-diphenyl-N,N′-bis(4-methylphenyl)-1,4-phenylenediamine can be condensed with paraldehyde to synthesize arylamine polymers for the fabrication of perovskite cells.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and characterization of triphenylamine derivatives by oxidative polymerization
Sim JH, et al.
Synthetic Metals, 157(22-23), 940-944 (2007)
Arylamine polymers prepared via facile paraldehyde addition condensation: an effective hole-transporting material for perovskite solar cells
Suwa K, et al.
Polymer International, 67(6), 670-674 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service