Merck
  • AMPK upregulates KCa2.3 channels and ameliorates endothelial dysfunction in diet-induced obese mice.
AMPK upregulates KCa2.3 channels and ameliorates endothelial dysfunction in diet-induced obese mice.
Biochemical pharmacology (2020-11-14)
Zheng-Da Pang, Yan Wang, Zheng Song, Gang She, Xiao-Zhen Ma, Xia Sun, Wei Wu, Bao-Chang Lai, Jiao Zhang, Yi Zhang, Xiao-Jun Du, John Y J Shyy, Xiu-Ling Deng
ABSTRACT
The opening of endothelial small-conductance calcium-activated potassium channels (KCa2.3) is essential for endothelium-dependent hyperpolarization (EDH), which predominantly occurs in small resistance arteries. Adenosine monophosphate-activated protein kinase (AMPK), an important metabolic regulator, has been implicated in regulating endothelial nitric oxide synthase activity. However, it was unclear whether AMPK regulated endothelial KCa2.3-mediated EDH-type vasodilation. Using bioinformatics analysis and myograph system, we investigated the regulation by AMPK of KCa2.3 in human umbilical vein endothelial cells (HUVECs) or mouse second-order mesenteric resistance arteries. In HUVECs, AMPK activation either by activators (AICAR, A769662 and MK-8722) or expression of the constitutively active form of AMPK significantly upregulated KCa2.3 expression. Such effects were abolished by AMPK inhibitor (compound C) or AMPK α1-/α2-siRNA, extracellular-signal-regulated-kinase 5 (ERK5) inhibitor (ERK5-IN-1), and specific siRNA to myocyte-enhancer factor 2 (MEF2) or krüppel-like factor 2/4 (KLF2/4). KCa2.3 expression was significantly reduced in mesenteric resistance arteries in AMPKα2 knockout mice when compared with littermate control mice. Furthermore, in high-fat diet fed mice, 2-week treatment with AICAR restored endothelial KCa2.3 expression in mesenteric resistance arteries with improved endothelial dysfunction. Our results demonstrate that activation of AMPK upregulates KCa2.3 channel expression through the ERK5-MEF2-KLF2/4 signaling pathway in vascular endothelium, which contributes to benefits through KCa2.3-mediated EDH-type vasodilation in mesenteric resistance arteries.
Sigma-Aldrich
TRAM-34, ≥98% (HPLC), solid
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
NS309, ≥98% (HPLC), solid