97609
MOSH/MOAH Retention Time Mix
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
multi-component solution
storage temp.
room temp
Related Categories
Other Notes
The certified values are from the following products: n-Undecane, n-Tridecane, n-Hexadecane, n-Eicosane.
The other values are informative values not within the accreditation of ISO17034 and ISO/IEC17025.
The other values are informative values not within the accreditation of ISO17034 and ISO/IEC17025.
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Analyte
Description
n-Undecane
n-Tridecane
n-Hexadecane
n-Eicosane
n-Decane
n-Tetracosane
n-Pentacosane
n-Pentatriacontane
n-Tetracontane
n-Pentacontane
See All (10)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
-4.0 °F - closed cup
Flash Point(C)
-20 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service