Skip to Content
Merck
All Photos(1)

Key Documents

90152

Supelco

N4,N4-Diethyl-2-methyl-1,4-benzenediamine hydrochloride

suitable for GC, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N2 · HCl
CAS Number:
Molecular Weight:
214.73
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

grade

analytical standard

Quality Level

Assay

≥98.0 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-30°C

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service