Skip to Content
Merck
All Photos(1)

Key Documents

41898

Supelco

Methylmalonyl-L-carnitine-(N-methyl-d3) lithium salt

analytical standard, ≥90.0% (Mixture of diastereomers, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16D3NO6 · xLi+
Molecular Weight:
264.29 (free acid basis)
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥90.0% (Mixture of diastereomers, HPLC)

optical activity

[α]/D -39.0±2.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99%

application(s)

clinical testing

storage temp.

−20°C

SMILES string

C[N+](C[C@H](OC(C(C)C(O)=O)=O)CC([O-])=O)(C)C([2H])([2H])[2H]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service