Skip to Content
Merck
All Photos(1)

Key Documents

24622-U

Supelco

Vacuum Sampling Pumps

Synonym(s):

Air Sampling Pumps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

description

Not UL listed; Not EX rated

technique(s)

whole air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Single 1-2 Liter Bags

Looking for similar products? Visit Product Comparison Guide

General description

The battery charger is not included with part number 24622-U. Please order part number 24643 separately.
Vacuum Sampling Pumps is a field sampling pump, used in active and whole air samplers.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service