Skip to Content
Merck
All Photos(7)

Documents

ZRB2441

Sigma-Aldrich

Anti-OPA1 Antibody, clone 1F15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Dynamin-like 120 kDa protein, mitochondrial, EC:3.6.5.5, Optic atrophy protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F15, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 111.63 kDa
observed mol wt ~100 kDa

purified by

using Protein A

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... OPA1(4976)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F15 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Optic atrophy protein 1 (OPA1). It targets an epitope within 22 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal region of human Optic atrophy protein 1 (OPA1).

Application

Quality Control Testing

Evaluated by Western Blotting in Rat kidney tissue lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected OPA1 in Rat kidney tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected OPA1 in C2C12 cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound OPA1 peptide with a KD of 1.5 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected OPA1 in Human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Dynamin-like 120 kDa protein, mitochondrial (UniProt: O60313; also known as EC:3.6.5.5, OPTIC atrophy protein 1) is encoded by the OPA1 (also known as KIAA0567) gene (Gene ID: 4976) in human. OPA1 is a single-pass mitochondrial membrane protein that is synthesized with a TransIT® peptide (aa 1-87), which is subsequently cleaved off to produce the active protein that contains a short mitochondrial matrix domain (aa 88-96), a transmembrane domain (aa 97-113), and a mitochondrial intermembrane domain (aa 114-960). It is detected at contacts sites between endoplasmic reticulum and mitochondrial membranes. It is highly expressed in the retina and is also detected in the brain, testis, heart, skeletal muscle, and other tissues. It serves as a dynamin-related GTPase and is shown to be essential for normal mitochondrial morphology. It regulates the equilibrium between mitochondrial fusion and mitochondrial fission. At least six different isoforms of OPA1 have been described that are produced by alternative splicing. Co-expression of isoform 1 with shorter alternative products is required for its optimal activity in promoting mitochondrial fusion. It binds lipid membranes enriched in negatively charged phospholipids and promotes membrane tubulation. Its intrinsic GTPase activity is reported to be low and is strongly increased by interaction with lipid membranes. OPA1 also plays a role in remodeling cristae and the release of cytochrome c during apoptosis. Proteolytic processing in response to intrinsic apoptotic signals may lead to disassembly of OPA1 oligomers and release of cytochrome c into the mitochondrial intermembrane space. Mutations in this gene have been associated with OPTIC atrophy type 1, which is a dominantly inherited OPTIC neuropathy resulting in progressive loss of visual acuity, leading in many cases to blindness. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Anand, R., et al. (2014). J. Cell Biol. 204(6); 919-929; BAN, T., et al. (2010). Hum. Mol. Genet. 19(11); 2113-2122; Song, Z., et al. (2007). J. Cell Biol. 178(5); 749-755).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
BAN is a trademark of Novozymes Corp.
TransIT is a registered trademark of Mirus Bio
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service