Key Documents
Z635049
Biohit Proline® single channel electronic pipettor
volume 10-500 μL
About This Item
Pricing and availability is not currently available.
description
without charger
manufacturer/tradename
Sartorius 710100
availability
not available in ROW, USA and Canada
volume
10-500 μL
General description
Sartorius Proline electronic pipettors of the Biohitfamily offer effortless and rapid pipetting with high levels of accuracy and precision. They offer a wide variety of pipetting operations, making the Proline range the complete solution for all pipetting needs. Proline pipettors are ergonomically designed and the tip ejection is easily operated with four fingers to avoid strain and fatigue during pipetting. A microprocessor-controlled piston ensures optimal precision and accuracy at the same time being user-independent, delivering reliable results consistently. Proline liquid handling operations include:
  • Forward Pipetting
  • Reverse Pipetting (excl. 1200, 5000)
  • Multiple-Dispensing
  • Multi-Aspirating (only 1200)
  • Diluting (excl. mcp 1200)
  • Sequential-Dispensing (only 1200, 5000)
  • Post-Delivery Mixing
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)