Skip to Content
Merck
All Photos(1)

Documents

Z363359

Sigma-Aldrich

Scienceware® multiwell plate tilt stand

can be tilted and locked in position with thumbwheels

Synonym(s):

plate stand

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.22

sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F18938-0000

color

none

General description

Clear acrylic platform can be tilted and locked in position with thumbwheels. Facilitates manual addition of reagents by making small volumes more visible; presents rows in tiers for easier access. Epoxy-coated heavy steel base provides stability.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service