Documents
TMO267556
Nunc® Edge 2.0 96-well Plate
untreated, polystyrene, sterile, without lid, clear, case of 160 ea
Synonym(s):
cell culture plate, cell growth plate
About This Item
UNSPSC Code:
41122102
NACRES:
NB.15
material
clear
polystyrene
sterility
sterile
packaging
case of 160 ea
manufacturer/tradename
Nunc 267556
General description
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific Nunc Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries