All Photos(1)
About This Item
Linear Formula:
CH3(CH2)21CH3
CAS Number:
Molecular Weight:
324.63
Beilstein:
1756000
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
~99%
bp
199-200 °C/3 mmHg (lit.)
mp
46-47 °C (lit.)
SMILES string
CCCCCCCCCCCCCCCCCCCCCCC
InChI
1S/C23H48/c1-3-5-7-9-11-13-15-17-19-21-23-22-20-18-16-14-12-10-8-6-4-2/h3-23H2,1-2H3
InChI key
FIGVVZUWCLSUEI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service