Skip to Content
Merck
All Photos(1)

Documents

SAB2108735

Sigma-Aldrich

Anti-TRIM72

affinity isolated antibody

Synonym(s):

Anti-MG53

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

horse, rat, human, bovine, pig

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

accession no.

NM_001008274

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM72(493829)

Immunogen

Synthetic peptide directed towards the middle region of human TRIM72

Biochem/physiol Actions

TRIM72 belongs to the TRIM/RBCC family. It contains 1 B box-type zinc finger and 1 B30.2/SPRY domain and 1 RING-type zinc finger. The function of TRIM72 remains unknown.

Sequence

Synthetic peptide located within the following region: LEELTFDPSSAHPSLVVSSSGRRVECSEQKAPPAGEDPRQFDKAVAVVAH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clothilde Philouze et al.
PloS one, 16(2), e0245179-e0245179 (2021-02-11)
In type 2 diabetes (T2D), both muscle and liver are severely resistant to insulin action. Muscle insulin resistance accounts for more than 80% of the impairment in total body glucose disposal in T2D patients and is often characterized by an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service