Skip to Content
Merck
All Photos(3)

Key Documents

SAB1412661

Sigma-Aldrich

ANTI-RXRA antibody produced in mouse

clone 3D9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ00280, FLJ00318, FLJ16020, FLJ16733, RXRA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D9, monoclonal

form

buffered aqueous solution

mol wt

antigen 43.89 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RXRA(6256)

General description

Retinoid X receptors (RXRs) and retinoic acid receptors (RARs), are nuclear receptors that mediate the biological effects of retinoids by their involvement in retinoic acid-mediated gene activation. These receptors exert their action by binding, as homodimers or heterodimers, to specific sequences in the promoters of target genes and regulating their transcription. The protein encoded by this gene is a member of the steroid and thyroid hormone receptor superfamily of transcriptional regulators. (provided by RefSeq)

Immunogen

RXRA (AAH07925, 1 a.a. ~ 165 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLGSWPARTFHPGACVSRRPSAPWKHTASGKDSPDLRFSEHGVSQEFWAGGLVAVLEMTPSPSPWGTQEGPAGMCSLWVVGWCPCRGAGVRDLVLVHAGVWCKHVCAVQRDACGESRTPAPPRKGGAVTSVLCLFLIKTFPLFSYKFASCKQVHKDPPLVKSGFE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service