Skip to Content
Merck
All Photos(2)

Key Documents

SAB1405444

Sigma-Aldrich

Anti-ALAD antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ALADH, MGC5057, PBGS

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
NOK 4,170.00

NOK 4,170.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
NOK 4,170.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

NOK 4,170.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~37.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALAD(210)

General description

The ALAD enzyme is composed of 8 identical subunits and catalyzes the condensation of 2 molecules of delta-aminolevulinate to form porphobilinogen (a precursor of heme, cytochromes and other hemoproteins). ALAD catalyzes the second step in the porphyrin and heme biosynthetic pathway; zinc is essential for enzymatic activity. ALAD enzymatic activity is inhibited by lead and a defect in the ALAD structural gene can cause increased sensitivity to lead poisoning and acute hepatic porphyria. (provided by RefSeq)

Immunogen

ALAD (NP_000022.2, 1 a.a. ~ 339 a.a) full-length human protein.

Sequence
MPLCPLAHAMQPQSVLHSGYFHPLLRAWQTATTTLNASNLIYPIFVTDVPDDIQPITSLPGVARYGVKRLEEMLRPLVEEGLRCVLIFGVPSRVPKDERGSAADSEESPAIEAIHLLRKTFPNLLVACDVCLCPYTSHGHCGLLSENGAFRAEESRQRLAEVALAYAKAGCQVVAPSDMMDGRVEAIKEALMAHGLGNRVSVMSYSAKFASCFYGPFRDAAKSSPAFGDRRCYQLPPGARGLALRAVDRDVREGADMLMVKPGMPYLDIVREVKDKHPDLPLAVYHVSGEFAMLWHGAQAGAFDLKAAVLEAMTAFRRAGADIIITYYTPQLLQWLKEE

Application

Anti-ALAD antibody produced in mouse is suitable for western blot application.

Biochem/physiol Actions

The cytosolic ALAD (δ-aminolevulinic acid dehydratase) plays a vital role in the heme biosynthetic pathway. It facilitates the second step, which is a condensation reaction of 5-aminolevulinic acid and forms a monopyrrole termed as porphobilinogen. It is one of the main targets of lead toxicity. Lead replaces zinc present in ALAD, leading to inactivation of the enzyme. Deficiency in ALAD causes ALAD deficiency porphyria (ADP).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E K Jaffe et al.
The Journal of biological chemistry, 276(2), 1531-1537 (2000-10-18)
Human porphobilinogen synthase (PBGS) is a main target in lead poisoning. Human PBGS purifies with eight Zn(II) per homo-octamer; four ZnA have predominantly nonsulfur ligands, and four ZnB have predominantly sulfur ligands. Only four Zn(II) are required for activity. To
N Ishida et al.
The Journal of clinical investigation, 89(5), 1431-1437 (1992-05-01)
Cloning and expression of the defective genes for delta-aminolevulinate dehydratase (ALAD) from a patient with inherited ALAD deficiency porphyria (ADP) were carried out. Cloning of cDNAs for the defective ALAD were performed from EBV-transformed lymphoblastoid cells of the proband, and
Christine Neslund-Dudas et al.
The Prostate, 74(6), 637-646 (2014-02-07)
Black men have historically had higher blood lead levels than white men in the U.S. and have the highest incidence of prostate cancer in the world. Inorganic lead has been classified as a probable human carcinogen. Lead (Pb) inhibits delta-aminolevulinic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service