PZ0262
PF-06663195
≥98% (HPLC)
Synonym(s):
(4S,4aR,6R,8aS)-8a-(2,4-Difluorophenyl)-4-(fluoromethyl)-4,4a,5,6,8,8a-hexahydro-6-(3-methyl-5-isoxazolyl)-pyrano[3,4-d][1,3]thiazin-2-amine, (4S,4aR,6R,8aS)-8a-(2,4-Difluorophenyl)-4-(fluoromethyl)-6-(3-methyl-1,2-oxazol-5-yl)-4,4a,5,6,8,8a-hexahydropyrano[3,4-d][1,3]thiazin-2-amine, PF 06663195, PF06663195
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 20 mg/mL, clear
storage temp.
room temp
SMILES string
[H][C@@]12[C@@](CO[C@@H](C3=CC(C)=NO3)C2)(C4=CC=C(F)C=C4F)N=C(N)S[C@@H]1CF
InChI
1S/C18H18F3N3O2S/c1-9-4-15(26-24-9)14-6-12-16(7-19)27-17(22)23-18(12,8-25-14)11-3-2-10(20)5-13(11)21/h2-5,12,14,16H,6-8H2,1H3,(H2,22,23)/t12-,14+,16+,18+/m0/s1
InChI key
JIAYOIBUKDISBO-IYPIGGPWSA-N
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service