P6861
Standard PCR microtubes
Rigid PCR plate (96 well, low profile)
All Photos(1)
About This Item
UNSPSC Code:
41121702
NACRES:
NB.22
material
natural
round bottom
sterility
non-sterile
manufacturer/tradename
Thermo Scientific AB-0700
technique(s)
PCR: suitable
capacity
0.2 mL
General description
Precision moulded from virgin polypropylene and clean room produced.
Certified free from DNase, RNase and human genomic DNA contamination.
Certified free from DNase, RNase and human genomic DNA contamination.
Skirted low-profile plate
Suitable for 0.2 mL thermal cycler blocks
0.2 mL maximum well volume (when used with adhesive and heat seals)
Raised well rims for effective sealing and reduced cross-contamination risk
Sealable using mats, films and foils or cap strips
Suitable for 0.2 mL thermal cycler blocks
0.2 mL maximum well volume (when used with adhesive and heat seals)
Raised well rims for effective sealing and reduced cross-contamination risk
Sealable using mats, films and foils or cap strips
Choose from one of the most recent versions:
Certificates of Analysis (COA)