MTCC2625R
MTC™ Bio Screw Cap Centrifuge Tube
capacity 25 mL, conical bottom, sterile, pack of 200 ea (8 x foam racks 25 ea)
Synonym(s):
conical tube, plastic tube
All Photos(1)
About This Item
Recommended Products
material
blue cap
clear tube
conical bottom
polypropylene
round
screw top cap
sterility
sterile
packaging
pack of 200 ea (8 x foam racks 25 ea)
manufacturer/tradename
MTC™ Bio C2625-R
parameter
12,000 × g speed (max)
capacity
25 mL
diam. × L
27 mm × 78 mm
Related Categories
General description
Centrifuge Tube, 25mL, PP, sterile, with screw cap, 8 foam racks of 25 tubes, 200/pack
These centrifuge tubes are made of premium polypropylene to maximize centrifugal strength and maintain clarity. Tubes and caps may be frozen at temperatures as low as 86˚C. Caps have a flat top surface for easy marking and labeling.
Features and Benefits
- Sterile, RNase, DNase, pyrogen and cytotoxin free
- Available racked or bagged
- Compatible with standard rotors
- Frosted marking surface
Legal Information
MTC is a trademark of MTC Bio