Skip to Content
Merck
All Photos(1)

Documents

HS10044H

Wire biohazard bag holder

W × L 91 mm × 128 mm, red

Synonym(s):

Biohazard Bag Stand

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24111500
NACRES:
NB.17

material

epoxy (coated steel)

feature

Holds 1 x bag

packaging

pack of 1 ea

manufacturer/tradename

Heathrow Scientific HS10044H

W × L

91 mm × 128 mm

color

red

General description

Epoxy coated steel wire holder for Biohazard Bags

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service