H1038
Hck human
recombinant, expressed in E. coli BL21, vial of ≥200 units
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
recombinant
expressed in E. coli BL21
Quality Level
form
solution
specific activity
1-2 unit/μg protein
packaging
vial of ≥200 units
relevant disease(s)
cancer (leukemia)
shipped in
dry ice
storage temp.
−70°C
Looking for similar products? Visit Product Comparison Guide
Application
Hck is a protein-tyrosine kinase that is expressed in hemopoietic cells. It is used in leukemia research .
Biochem/physiol Actions
Hck is a member of the Src tyrosine kinase family. Hck has been shown to associate with and be activated by Bcr-Abl, which is a protein-tyrosine kinase that is expressed in chronic myelogenous leukemia .
Unit Definition
One unit will phosphorylate 1.0 picomole of Tyr15 of the peptide substrate pcdc2 6-20 per minute at 30 °C at pH 7.4.
Physical form
Supplied as a solution in 50 mM Tris HCl, pH 7.4, 1 mM EDTA and 1 mM DTT
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of biological chemistry, 275(24), 18581-18585 (2000-06-13)
Bcr-Abl is the constitutively active protein-tyrosine kinase expressed as a result of the Philadelphia translocation in chronic myelogenous leukemia. Bcr-Abl is coupled to many of the same signaling pathways normally regulated by hematopoietic cytokines. Recent work shows that Hck, a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service