G0157
Anti-GABA Transporter GAT-1 antibody produced in rabbit
affinity isolated antibody, lyophilized powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352203
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
rat
technique(s)
immunohistochemistry: suitable
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Slc6a12(50676)
Immunogen
synthetic peptide corresponding to the C-terminal domain (amino acids 588-599) of rat GAT-1.
Physical form
Supplied as a lyophilized powder containing 1% bovine serum albumin.
Not finding the right product?
Try our Product Selector Tool.
comparable product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chitoshi Takayama et al.
Brain research. Developmental brain research, 158(1-2), 41-49 (2005-07-19)
In the brain, gamma-amino butyric acid (GABA), released extrasynaptically and synaptically from GABAergic neurons, plays important roles in morphogenesis, expression of higher functions and so on. In the GABAergic transmission system, plasma membrane GABA transporters (GATs) mediate GABA-uptake from the
Xingwu Zhong et al.
Investigative ophthalmology & visual science, 45(7), 2065-2074 (2004-06-30)
To demonstrate regulation of ocular growth and refractive development by image quality and processing in the retina in higher primates. Focus-sensitive retinal neurons were labeled with immunocytochemical markers after briefly altering image quality in infant monkeys. Six rhesus monkeys (Macaca
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service