Skip to Content
Merck
All Photos(1)

Documents

EP5820740000

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

S-4-104, for 5810/5810R, aerosol-tight, with 4 x 750 mL round buckets

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

feature

to hold 4 x 750 mL round buckets
aerosol-tight
for 5810/5810R
high-capacity swing-bucket rotor
with 4 x 750 mL round buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5820740000 (Global)

parameter

3,214 × g max. RCF
3,900 rpm

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

Looking for similar products? Visit Product Comparison Guide

General description

  • Medium-capacity swing bucket rotor holds tubes and bottles from 0.2 mL to 750 mL as well as plates.
  • Rotor with 4 × 750 mL round buckets features a QuickLock rotor lid for ergonomic lid locking.
  • The aerosol-tight cap facilitates safe centrifugation of potentially hazardous materials.
  • Mixed loading with 2 plate buckets and 2 round tube buckets allows for increased versatility.
  • Unique aerodynamic round bucket design aids in saving energy up to 36% compared to Rotor A-4-81.
  • Buckets, lids and adapters are autoclavable (121 °C, 20 min).
  • The S-4-104 rotor is not compatible with the 5804/5804R centrifuge. It can only be used with the 5810/5810R centrifuge.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service