EP5804727002
Eppendorf® Fixed-angle rotor
F-34-6-38, 6 x 85 ml incl. lid
Synonym(s):
centrifuge laboratory, lab centrifuge, micro centrifuge
About This Item
Recommended Products
feature
to hold 6 x 85 mL tubes
F-34-6-38, 6 x 85 ml incl. lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5804727002
parameter
12,000 (5810R), 11,000 (5804/5804R/5810) max. RCF (18,514 xg (5810R), 15,557 xg (5804/5804R/5810))
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
Looking for similar products? Visit Product Comparison Guide
General description
- A fixed-angle rotor (F-34-6-38) is ideal for high-speed centrifugation of samples for molecular applications.
- The rotor is lightweight that facilitates convenient and easy handling.
- Made of chemically resistant anodized aluminum.
- Rotor, lid and adapters are autoclavable (121 °C, 20 min).
- Features a 34° angle borehole to prevent pellet smears along tube wall.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service