Skip to Content
Merck
All Photos(1)

Documents

CLS7248

Corning® TC treated 1536 well microplates

black, with lid, sterile

Synonym(s):

96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, EIA/RIA plate, immunoassay plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

black
black bottom
polystyrene plate
square wells

sterility

sterile

feature

lid
skirt

packaging

case of 50 ea

manufacturer/tradename

Corning 7248

size

1536 wells

well volume

12.5 μL

well working volume

1-8 μL

color

black

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Superior performance compared to other available commercial microplates: lower CV values, excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Optimized for automation with bar coding and standard readers
  • Surface treated for optimal cell attachment
  • Flat bottom wells with lid are compatible with automated dispensers including the PerkinElmer® FlexDrop
  • Sterile and certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.
PerkinElmer is a registered trademark of PerkinElmer Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service