Documents
CLS3584
Corning® volume adapter for 96 well filter plates
metal, non-sterile
About This Item
UNSPSC Code:
41121800
NACRES:
NB.24
material
metal
sterility
non-sterile
packaging
pack of 10 ea
case of 50 ea
manufacturer/tradename
Corning 3584
well volume
1 mL
wells
96 wells
General description
Volume adapter mounts on top of the 96 well FiltrEX~; filter plate and allows larger volumes (1mL) to be applied to each well in a filter plate. Adapters are nonsterile.
Legal Information
Corning is a registered trademark of Corning, Inc.
FiltrEX is a trademark of Corning, Inc.