CLS356640
Corning® BioCoat™ Poly-D-Lysine Microplate
well size 96 wells, flat bottom clear black wells
Synonym(s):
Cell Culture Plate, plastic coated cell culture plate
About This Item
material
flat bottom clear black wells
feature
lid: no
packaging
pkg of 50 ea (10 packs of 50)
manufacturer/tradename
Corning 356640
well size
96 wells
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service